MilliporeSigma
  • The relationship between the plasma concentration of blonanserin, and its plasma anti-serotonin 5-HT(2A) activity/anti-dopamine D₂ activity ratio and drug-induced extrapyramidal symptoms.
The relationship between the plasma concentration of blonanserin, and its plasma anti-serotonin 5-HT(2A) activity/anti-dopamine D₂ activity ratio and drug-induced extrapyramidal symptoms.
Psychiatry and clinical neurosciences (2012-02-23)
Hidenobu Suzuki, Keishi Gen
ABSTRACT
Blonanserin is a second-generation antipsychotic that was developed in Japan. We investigated the relationships between plasma concentration, the plasma anti-5-HT(2A) activity/anti-D₂ activity (S/D) ratio and extrapyramidal symptoms (EPS) in blonanserin dosing. The subjects were 29 outpatients with schizophrenia. We assessed EPS using the Drug-Induced Extrapyramidal Symptoms Scale (DIEPSS). The plasma concentrations were measured by high performance liquid chromatography, and the plasma anti-D₂ and anti-5-HT(2A) activities were measured by [³H]-spiperone and [³H]-ketanserin radioreceptor assays. The results revealed that there were significant correlations between both the plasma concentration and the DIEPSS total score (P<0.05). A negative correlative tendency was found between the S/D ratio and the DIEPSS total score. Furthermore, the plasma concentrations were divided into a low plasma concentration group and a high plasma concentration group, and the S/D ratios were divided into a low S/D ratio group and a high S/D ratio group. We then compared each group based on the DIEPSS total scores. The score in the high plasma concentration-low S/D ratio group was significantly higher than in the high plasma concentration-high S/D ratio, low plasma concentration-high S/D ratio and low plasma concentration-low S/D ratio groups (P<0.05 for all). These findings indicate that the incidence of EPS during treatment with blonanserin is mainly determined by plasma concentration, but the incidence of EPS may be inhibited when anti-5HT(2A) activity is predominant over anti-D₂ activity.
Sigma-Aldrich
Blonanserin, ≥98% (HPLC)