MilliporeSigma
Follow-up of natural products isolation.
Methods in molecular biology (Clifton, N.J.) (2012-03-01)
Richard J P Cannell, Satyajit D Sarker, Lutfun Nahar
ABSTRACT
Follow-up of natural products isolation refers to re-isolation of compound(s) of interest in larger amounts for further pharmacological testing, conclusive structure elucidation, structure modifications to synthesize analogs for structure-activity relationships (SAR) studies, preformulation and formulation studies or clinical trials. In addition to conventional synthetic chemistry approaches, several other methodologies can be applied for following-up natural products isolation. This chapter outlines, with specific examples, various strategies and methods involved in follow-up of natural products isolation.
Sigma-Aldrich
Nonactin, from Streptomyces griseus, ≥98.0% (Total homologs, HPLC)
Sigma-Aldrich
Ammonium ionophore I, Selectophore, function tested, ≥98.0% ((Total homologs), HPLC)