MilliporeSigma
  • Quality changes of sea bass slices wrapped with gelatin film incorporated with lemongrass essential oil.
Quality changes of sea bass slices wrapped with gelatin film incorporated with lemongrass essential oil.
International journal of food microbiology (2012-03-01)
Mehraj Ahmad, Soottawat Benjakul, Punnanee Sumpavapol, Nilesh Prakash Nirmal
ABSTRACT
Microbiological, chemical and physical changes of sea bass slices wrapped with gelatin film incorporated with 25% (w/w) lemongrass essential oil (LEO) during storage of 12 days at 4 °C were investigated. Sea bass slices wrapped with LEO film had the retarded growth of lactic acid bacteria (LAB), psychrophilic bacteria and spoilage microorganisms including H₂S-producing bacteria and Enterobacteriaceae throughout storage of 12 days in comparison with the control and those wrapped with gelatin film without LEO (G film) (P<0.05). Lowered changes of colour, K value, total volatile base nitrogen (TVB) and TBARS value were also found in LEO film wrapped samples, compared with those wrapped with G film and control, respectively. Therefore, the incorporation of LEO into gelatin film could enhance the antimicrobial and antioxidative properties of the film, thereby maintaining the qualities and extending the shelf-life of the sea bass slices stored at refrigerated temperature.