MilliporeSigma
  • A comparison of the effects of symmetry and magnetoanisotropy on paramagnetic relaxation in related dysprosium single ion magnets.
A comparison of the effects of symmetry and magnetoanisotropy on paramagnetic relaxation in related dysprosium single ion magnets.
Chemical communications (Cambridge, England) (2012-04-26)
Ursula J Williams, Brian D Mahoney, Patrick T DeGregorio, Patrick J Carroll, Eiko Nakamaru-Ogiso, James M Kikkawa, Eric J Schelter
ABSTRACT
Dysprosium complexes of the tmtaa(2-) ligand were synthesized and characterized by X-band EPR and magnetism studies. Both complexes demonstrate magnetoanisotropy and slow paramagnetic relaxation. Comparison of these compounds with the seminal phthalocyanine complex [Dy(Pc)(2)](-) shows the azaannulide complexes are more susceptible to relaxation through non-thermal pathways.
Sigma-Aldrich
Dysprosium, chips, 99.9% trace rare earth metals basis
Sigma-Aldrich
Dysprosium, ingot, 99.9% trace rare earth metals basis