MilliporeSigma
  • Blonanserin, a novel atypical antipsychotic agent not actively transported as substrate by P-glycoprotein.
Blonanserin, a novel atypical antipsychotic agent not actively transported as substrate by P-glycoprotein.
Progress in neuro-psychopharmacology & biological psychiatry (2012-06-14)
Tomoko Inoue, Kenichi Osada, Masaaki Tagawa, Yuriko Ogawa, Toshiaki Haga, Yoshihisa Sogame, Takanori Hashizume, Takashi Watanabe, Atsushi Taguchi, Takashi Katsumata, Masashi Yabuki, Noboru Yamaguchi
ABSTRACT
Although blonanserin, a novel atypical antipsychotic agent with dopamine D(2)/serotonin 5-HT(2A) antagonistic properties, displays good brain distribution, the mechanism of this distribution has not been clarified. P-glycoprotein [(P-gp) or multidrug resistance protein 1 (MDR1)] is an efflux transporter expressed in the brain and plays an important role in limiting drug entry into the central nervous system (CNS). In particular, P-gp can affect the pharmacokinetics and efficacy of antipsychotics, and exacerbate or soothe their adverse effects. In this study, we conducted in vitro and in vivo experiments to determine whether blonanserin is a P-gp substrate. Risperidone and its active metabolite 9-hydroxyrisperidone, both of which are P-gp substrates, were used as reference drugs. Affinity of blonanserin, risperidone, and 9-hydroxyrisperidone for P-gp was evaluated by in vitro transcellular transport across LLC-PK1, human MDR1 cDNA-transfected LLC-PK1 (LLC-MDR1), and mouse Mdr1a cDNA-transfected LLC-PK1 (LLC-Mdr1a). In addition, pharmacokinetic parameters in the brain and plasma (B/P ratio) of test compounds were measured in mdr1a/1b knockout (KO) and wild-type (WT) mice. The results of in vitro experiments revealed that P-gp does not actively transport blonanserin as a substrate in humans or mice. In addition, blonanserin displayed comparable B/P ratios in KO and WT mice, whereas B/P ratios of risperidone and 9-hydroxyrisperidone differed markedly in these animals. Our results indicate that blonanserin is not a P-gp substrate and therefore its brain distribution is unlikely to be affected by this transporter.
Sigma-Aldrich
Blonanserin, ≥98% (HPLC)