MilliporeSigma
  • Determination of benazolin-ethyl residues in soil and rape seed by SPE clean-up and GC with electron capture detection.
Determination of benazolin-ethyl residues in soil and rape seed by SPE clean-up and GC with electron capture detection.
Journal of chromatographic science (2012-06-22)
Xiaolu Liu, Tao Yang, Jiye Hu
ABSTRACT
A method has been developed and established for residue determination of benazolin-ethyl in soil and rape seed samples by gas chromatography with electron capture detection (GC-ECD). Limits of quantification of the method are 0.005 mg/kg for both soil and rape seed, which are sufficiently below the maximum residue limit, and the limit of detection is 0.0023 ng. The average recoveries of the analyte range from 85.89 to 105.84% with relative standard deviations (coefficient of variation) less than 5.53% at the three spike levels (0.005, 0.1 and 0.5 mg/kg). The half-life of benazolin-ethyl in soil from the experimental field is 4.62 days. The final residues of benazolin-ethyl in soil and rape seed samples are lower than 0.005 mg/kg at harvest time. Direct confirmation of the analyte in real samples is achieved by GC-mass spectrometry. It is demonstrated that the proposed method is simple, rapid and efficient, and reliable to detect benazolin-ethyl residues in soil and rape seed samples.