MilliporeSigma
  • [Reversible lupininin inhibitors of cholinesterases of mammalian blood and of optical ganglia of individuals of the commander squid Berryteuthis magister from different zones of species areal].
[Reversible lupininin inhibitors of cholinesterases of mammalian blood and of optical ganglia of individuals of the commander squid Berryteuthis magister from different zones of species areal].
Zhurnal evoliutsionnoi biokhimii i fiziologii (2012-07-26)
N E Basova, B N Kormilitsyn, A Iu Perchenok, E V Rozengart, V S Saakov, A A Suvorov
ABSTRACT
Arylsulfoesters and carbonic lupinin esters are studied for the first time as reversible inhibitors of mammalian blood cholinesterases. Studied in detail is sensitivity of cholinesterases to mono- and bislupinin inhibitors in Commander squid individuals from different habitation zones.