MilliporeSigma
  • Influence of ageing treatment on microstructure, mechanical and bio-corrosion properties of Mg-Dy alloys.
Influence of ageing treatment on microstructure, mechanical and bio-corrosion properties of Mg-Dy alloys.
Journal of the mechanical behavior of biomedical materials (2012-07-31)
Lei Yang, Yuanding Huang, Frank Feyerabend, Regine Willumeit, Karl Ulrich Kainer, Norbert Hort
ABSTRACT
Mg-Dy alloys have shown to be promising for medical applications. In order to investigate the influence of ageing treatment on their mechanical and corrosion properties, three Mg-xDy alloys (x=10, 15, 20 wt%) were prepared. Their microstructure, mechanical and corrosion behavior were investigated. The results indicate that ageing at 250 °C has little influence on the mechanical and corrosion properties. In contrast, ageing at 200 °C significantly increases the yield strength, and reduces the ductility. After ageing at 200 °C, the corrosion rate of Mg-20Dy alloy increases largely in 0.9 wt% NaCl solution, but remains unchanged in cell culture medium.
Sigma-Aldrich
Dysprosium, ingot, 99.9% trace rare earth metals basis
Sigma-Aldrich
Dysprosium, chips, 99.9% trace rare earth metals basis