MilliporeSigma
Total synthesis of pentosidine.
Organic letters (2012-08-23)
Adam J Rosenberg, Daniel A Clark
ABSTRACT
Pentosidine, a biologically important advanced glycation endproduct, has been accessed in a rapid, high-yielding manner. The synthesis was accomplished via a six-step sequence starting with 3-amino-2-chloropyridine and features a palladium-catalyzed tandem cross-coupling/cyclization to construct the imidazo[4,5-b]pyridine core.
Sigma-Aldrich
4-Azabenzimidazole, 99%