MilliporeSigma
  • Endoplasmic Reticulum stress induces hepatic stellate cell apoptosis and contributes to fibrosis resolution.
Endoplasmic Reticulum stress induces hepatic stellate cell apoptosis and contributes to fibrosis resolution.
Liver international : official journal of the International Association for the Study of the Liver (2012-09-04)
Samuele De Minicis, Cinzia Candelaresi, Laura Agostinelli, Silvia Taffetani, Stefania Saccomanno, Chiara Rychlicki, Luciano Trozzi, Marco Marzioni, Antonio Benedetti, Gianluca Svegliati-Baroni
ABSTRACT
Survival of hepatic stellate cells (HSCs) is a hallmark of liver fibrosis, while the induction of HSC apoptosis may induce recovery. Activated HSC are resistant to many pro-apoptotic stimuli. To this issue, the role of Endoplasmic Reticulum (ER) stress in promoting apoptosis of HSCs and consequently fibrosis resolution is still debated. To evaluate the potential ER stress-mediated apoptosis of HSCs and fibrosis resolution HSCs were incubated with the ER stress agonists, tunicamycin or thapsigargin. In vivo, HSC were isolated from normal, bile duct-ligated (BDL) and bile duct-diverted (BDD) rats. In activated HSC, the specific inhibitor of ER stress-induced apoptosis, calpastatin, is significantly increased vs. quiescent HSCs. Calpain is conversely reduced in activated HSCs. This pattern of protein expression provides HSCs resistance to the ER stress signals of apoptosis (apoptosis-resistant phenotype). However, both tunicamycin and thapsigargin are able to induce apoptosis in HSCs in vitro, completely reversing the calpain/calpastatin pattern expression. Furthermore, in vivo, the fibrosis resolution observed in rat livers subjected to bile duct ligation (BDL) and subsequent bile duct diversion (BDD), leads to fibrosis resolution through a mechanism of HSCs apoptosis, potentially associated with ER stress: in fact, BDD rat liver shows an increased number of apoptotic HSCs associated with reduced calapstatin and increased calpain protein expression, leading to an apoptosis-sensible phenotype. ER stress sensitizes HSC to apoptosis both in vitro and in vivo. Thus, ER stress represents a key target to trigger cell death in activated HSC and promotes fibrosis resolution.