MilliporeSigma
  • Evaluation of the endogenous cannabinoid system in mediating the behavioral effects of dipyrone (metamizol) in mice.
Evaluation of the endogenous cannabinoid system in mediating the behavioral effects of dipyrone (metamizol) in mice.
Behavioural pharmacology (2012-09-08)
Joel E Schlosburg, Lilyana Radanova, Vincenzo Di Marzo, Peter Imming, Aron H Lichtman
ABSTRACT
Dipyrone is a common nonopioid analgesic and antipyretic, which, in many countries, is available over the counter and is more widely used than paracetamol or aspirin. However, the exact mechanisms by which dipyrone acts remain inconclusive. Two novel arachidonoyl-conjugated metabolites are formed in mice following the administration of dipyrone that are dependent on the activity of fatty acid amide hydrolase (FAAH), which also represents the major catabolic enzyme of the endogenous cannabinoid ligand anandamide. These arachidonoyl metabolites not only inhibit cyclooxygenase (COX-1/COX-2) but also bind to cannabinoid receptors at low micromolar concentrations. The relative contributions of cannabinoid receptors and FAAH in the overall behavioral response to dipyrone remain untested. Accordingly, the two primary objectives of the present study were to determine whether the behavioral effects of dipyrone would (a) be blocked by cannabinoid receptor antagonists and (b) occur in FAAH mice. Here, we report that thermal antinociceptive, hypothermic, and locomotor suppressive actions of dipyrone are mediated by a noncannabinoid receptor mechanism of action and occurred after acute or repeated administration irrespective of FAAH. These findings indicate that FAAH-dependent arachidonoyl metabolites and cannabinoid receptors are not requisites by which dipyrone exerts these pharmacological effects under noninflammatory conditions.