MilliporeSigma
  • The interactions between piperonyl butoxide and E4, a resistance-associated esterase from the peach-potato aphid, Myzus persicae Sulzer (Hemiptera: Aphididae).
The interactions between piperonyl butoxide and E4, a resistance-associated esterase from the peach-potato aphid, Myzus persicae Sulzer (Hemiptera: Aphididae).
Pest management science (2012-09-13)
Despina Philippou, Valerio Borzatta, Elisa Capparella, Leni Moroni, Linda Field, Graham Moores
ABSTRACT
It has been reported previously that piperonyl butoxide (PBO) can inhibit both P450 and esterase activity. Although the method by which PBO combines with cytochrome P450 has been identified, the way in which it acts as an esterase inhibitor has not been established. This paper characterises the interactions between PBO and the resistance-associated esterase in Myzus persicae, E4. After incubation with PBO/analogues, hydrolysis of 1-naphthyl acetate by E4 is increased, but sequestration of azamethiphos is reduced. Rudimentary in silico modelling suggests PBO docks at the lip of the aromatic gorge. PBO binds with E4 to accelerate small substrates to the active-site triad, while acting as a blockade to larger, insecticidal molecules. Structure-activity studies with analogues of PBO also reveal the essential chemical moieties present in the molecule.
Sigma-Aldrich
Piperonylbutoxide, technical grade, 90%
Supelco
Piperonylbutoxide, PESTANAL®, analytical standard