MilliporeSigma
  • Protein kinase Cα/β inhibitor Gö6976 promotes PC12 cell adhesion and spreading through membrane recruitment and activation of protein kinase Cδ.
Protein kinase Cα/β inhibitor Gö6976 promotes PC12 cell adhesion and spreading through membrane recruitment and activation of protein kinase Cδ.
Experimental cell research (2012-10-16)
Sung Youn Jung, O Bok Kim, Hyun Ki Kang, Da Hyun Jang, Byung-Moo Min, Frank H Yu
ABSTRACT
Gö6976 is a nonglycosidic indolocarbazole compound widely used as a specific inhibitor of PKCα/β. In experiments probing for a role of PKCα in human laminin-2-integrin-mediated cell adhesion and spreading of PC12 cells, we observed unexpected enhancements of adhesion, spreading and stress fiber formation to 1 μM Gö6976 with concomitant increase in membrane translocation of PKCδ and autophosphorylation of focal adhesion kinase (FAK). Importantly, enhanced cellular behavior and membrane translocation of PKCδ induced by Gö6976 was retained in siRNA-transfected PC12 cells to knockdown PKCα expression. Gö6976 also induced laminin-dependent cell adhesion in NIH/3T3 and CV-1 fibroblasts, suggesting of a mechanism that may be common to multiple cell-types. A specific inhibitor of PKCδ, rottlerin, completely abrogated Gö6976-dependent increase in PC12 cell adhesion to laminin as well as the activation of small GTPases, Rac1 and Cdc42, that are downstream of PKCδ in adhesion receptor signaling. siRNA knockdown of Rac1 and Cdc42 expression inhibited cell spreading and lamellipodia formation in PC12 cells. Overall, these results suggest that Gö6976 may stimulate membrane recruitment of PKCδ through a mechanism that is independent of PKCα/β signaling. In addition, the activation of Rac1 and Cdc42 by human laminin-2-integrin-dependent activation of PKCδ/FAK signaling mediates cell spreading and lamellipodia formation in PC12 cells.