MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
(-)-Carvone: antispasmodic effect and mode of action.
Fitoterapia (2012-10-30)
Fábia Valéria M Souza, Marcelly Barbosa da Rocha, Damião P de Souza, Rosilene Moretti Marçal
ABSTRACT
(-)-Carvone is a monoterpene ketone found in spearmint (Mentha spicata var. crispa) essential oil that is widely used as an odor and flavor additive. An intestinal antispasmodic effect was recently reported for (-)-carvone, and it has been shown to be more potent than its (+)-antipode. The mechanism of (-)-carvone action in the intestines has not been investigated. To gain a better understanding of the (-)-carvone antispasmodic effect, we investigated its pharmacological effects in the guinea pig ileum. Terminal portions of the ileum were mounted for isotonic contraction recordings. The effect of (-)-carvone was compared with that of the classical calcium channel blocker (CCB) verapamil. In isolated ileal smooth muscle, (-)-carvone did not produce direct contractile or relaxation responses and did not modify electrically elicited contractions or low K(+)-evoked contractions. The submaximal contractions induced by histamine (p<0.001), BaCl2 (p<0.05), and carbachol (p<0.01) were significantly reduced by (-)-carvone. The contractile response elicited by high concentrations of carbachol was reduced but not abolished by (-)-carvone. No additive action was detected with co-incubation of (-)-carvone and verapamil on carbachol-induced contraction. (-)-Carvone reduced the contraction induced by high K(+) and was almost 100 times more potent than verapamil. Thus, (-)-carvone showed a typical and potent CCB-like action. Many effects described for both (-)-carvone and spearmint oil can be explained as a CCB-like mode of action.
Sigma-Aldrich
Barium chloride dihydrate, ≥99%
Sigma-Aldrich
Barium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Barium chloride dihydrate, ≥99.999% trace metals basis
Supelco
(+)-Carvone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
D-Carvone, ≥96%, FG
Sigma-Aldrich
(S)-(+)-Carvone, 96%
Sigma-Aldrich
Barium chloride, AnhydroBeads, −10 mesh, 99.95% trace metals basis
Sigma-Aldrich
Barium chloride, 99.9% trace metals basis
Sigma-Aldrich
Barium chloride, 99.999% trace metals basis
Sigma-Aldrich
Barium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
(+)-Carvone, analytical standard