MilliporeSigma
  • Toxicological and histopathological effects of hydramethylnon on Atta sexdens rubropilosa (Hymenoptera: Formicidae) workers.
Toxicological and histopathological effects of hydramethylnon on Atta sexdens rubropilosa (Hymenoptera: Formicidae) workers.
Micron (Oxford, England : 1993) (2012-11-15)
Pâmela Decio, Elaine C M Silva-Zacarin, Fabiana C Bueno, Odair C Bueno
ABSTRACT
The leaf-cut ants are important agricultural pest, because they can cause intense defoliation in plants and destroy large areas cultivated. Although there are several works for the control of these insects by examining the toxicity of natural chemical compounds on various species of ants, few are focused on analyses of morphological changes caused in the affected organs. The aim of this study was to evaluate the effects of hydramethylnon on Atta sexdens rubropilosa workers through toxicological bioassays and morphological analysis of the post-pharyngeal glands, midgut, and Malpighian tubules of these ants. Hydramethylnon dissolved either in acetone (HA) or in a mixture of acetone and soy oil (HAO) was added to the artificial diet at a concentration of 200μg/mL. The workers fed daily with the diet containing hydramethylnon showed higher mortality than the controls, especially when HAO was used. Moreover, light and electron microscopy revealed morphological alterations in the midgut and Malpighian tubules of workers treated with HA, whereas alterations of the post-pharyngeal glands were observed in the HAO-treated group. These results indicated that the presence of soy oil provided an alternate route for the ingestion of the formicide's active ingredient and corroborated previous studies that suggested a role for the post-pharyngeal glands in lipid metabolism. Our findings suggest that the oil may carry hydramethylnon to the gland lumen, resulting in lower quantity of the active ingredient in the intestinal lumen and Malpighian tubules that explains the lower degree of morphological alterations in these structures in the workers treated with HAO. These results may provide insight into the toxicological effects of hydramethylnon on leaf-cutting ants and the use of vegetable oil as an adjuvant in baits to control ants.
Product Description
Supelco
Hydramethylnon, PESTANAL®, analytical standard