MilliporeSigma
  • Phosphine-catalyzed synthesis of 3,3-spirocyclopenteneoxindoles from γ-substituted allenoates: systematic studies and targeted applications.
Phosphine-catalyzed synthesis of 3,3-spirocyclopenteneoxindoles from γ-substituted allenoates: systematic studies and targeted applications.
The Journal of organic chemistry (2013-01-25)
Catherine Gomez, Maxime Gicquel, Jean-Christophe Carry, Laurent Schio, Pascal Retailleau, Arnaud Voituriez, Angela Marinetti
ABSTRACT
The phosphine-promoted [3 + 2] cyclizations between γ-substituted allenoates and arylideneoxindoles have been applied to the stereoselective synthesis of spiro(cyclopentene)oxindoles with trisubstituted cyclopentene units. It has been demonstrated that PPh(3) operates a very efficient control of the relative stereochemistry of the three stereogenic centers of the final spiranic products. Focused experiments have been carried out then so as to access carbocyclic analogues of an important series of anticancer agents inhibiting MDM2-p53 interactions.