MilliporeSigma
Contact dermatitis from Naphthol AS.
Contact dermatitis (1990-03-01)
J Roed-Petersen, W Batsberg, E Larsen
ABSTRACT
Naphthol AS, a coupling agent in cotton dyeing, caused allergic contact dermatitis in 5 patients, sensitized from wearing a special kind of shirt. Patch tests with Naphthol AS and pieces of shirt were positive in all 5 patients. Analysis of 1 of the shirts revealed the presence of Naphthol AS. 1 patient with hand dermatitis and positive patch tests to Naphthol AS and nickel was a needlewoman, probably sensitized to Naphthol AS from occupational contact with clothes.