MilliporeSigma
  • Mobilization of progenitor cells into peripheral blood by gamma-tocotrienol: a promising radiation countermeasure.
Mobilization of progenitor cells into peripheral blood by gamma-tocotrienol: a promising radiation countermeasure.
International immunopharmacology (2013-02-19)
Sugata Ray, Shilpa S Kulkarni, Kushal Chakraborty, Roli Pessu, Martin Hauer-Jensen, K Sree Kumar, Sanchita P Ghosh
ABSTRACT
Gamma-tocotrienol (GT3), a vitamin E isoform, is shown to induce high levels of granulocyte colony stimulating factor (G-CSF) in mice. G-CSF is a key cytokine used for stimulation of hematopoiesis, and mobilization of hematopoietic stem and progenitor cells into peripheral blood. GT3 is also shown to induce vascular endothelial growth factor (VEGF), another important cytokine necessary for vasculogenesis and endothelial progenitor mobilization. Since GT3 induces both these cytokines, we tested whether GT3 mobilizes hematopoietic and endothelial progenitors in mice. GT3 (200mg/kg) was injected in 10-week-old CD2F1 mice and mobilization of progenitors in peripheral blood was analyzed at 24, 48, and 72 h post-administration. Circulating hematopoietic progenitor cells (HPCs, Lin(-), cKit(+)), endothelial progenitor cells (EPCs, Lin(-), CD34(+), Flk(+)), and stromal progenitor cells (SPCs, Lin(-), CD29(+), CD105(+)) in peripheral blood mononuclear cells (PBMCs) were analyzed simultaneously by flow cytometry. Mobilized HPCs, EPCs and SPCs in PBMC were also measured by colony-forming unit (CFU) assay in progenitor-specific media. Three groups of mice received vehicle, GT3 and GT3 plus AMD3100, a receptor antagonist used to enhance mobilization. GT3 induced significant mobilization of all three progenitor cell types compared to vehicle in peripheral blood; AMD3100 enhanced GT3-induced mobilization even further. Mobilization of progenitor cells in peripheral blood by GT3 indicates that GT3 can be used as an alternative to G-CSF and VGEF to mobilize HPCs and EPCs.
Product Description
Supelco
γ-Tocotrienol, analytical standard