MilliporeSigma
  • DHFR 19-bp deletion and SHMT C1420T polymorphisms and metabolite concentrations of the folate pathway in individuals with Down syndrome.
DHFR 19-bp deletion and SHMT C1420T polymorphisms and metabolite concentrations of the folate pathway in individuals with Down syndrome.
Genetic testing and molecular biomarkers (2013-02-21)
Cristiani Cortez Mendes, Aline Maria Zanchetta de Aquino Raimundo, Luciana Dutra Oliveira, Bruna Lancia Zampieri, Gustavo Henrique Marucci, Joice Matos Biselli, Eny Maria Goloni-Bertollo, Marcos Nogueira Eberlin, Renato Haddad, Maria Francesca Riccio, Hélio Vannucchi, Valdemir Melechco Carvalho, Érika Cristina Pavarino
ABSTRACT
Down syndrome (DS) results from the presence and expression of three copies of the genes located on chromosome 21. Studies have shown that, in addition to overexpression of the Cystathionine β-synthase (CBS) gene, polymorphisms in genes involved in folate/homocysteine (Hcy) metabolism may also influence the concentrations of metabolites of this pathway. Investigate the association between Dihydrofolate reductase (DHFR) 19-base pair (bp) deletion and Serine hydroxymethyltransferase (SHMT) C1420T polymorphisms and serum folate and plasma Hcy and methylmalonic acid (MMA) concentrations in 85 individuals with DS. Molecular analysis of the DHFR 19-bp deletion and SHMT C1420T polymorphisms was performed by polymerase chain reaction (PCR) by difference in the size of fragments and real-time PCR allelic discrimination, respectively. Serum folate was quantified by chemiluminescence and plasma Hcy and MMA by liquid chromatography-tandem mass spectrometry. Individuals with DHFR DD/SHMT TT genotypes presented increased folate concentrations (p=0.004) and the DHFR II/SHMT TT genotypes were associated with increased MMA concentrations (p=0.008). In addition, the MMA concentrations were negatively associated with age (p=0.04). There is an association between DHFR DD/SHMT TT and DHFR II/SHMT TT combined genotypes and folate and MMA concentrations in individuals with DS.
Sigma-Aldrich
Methylmalonic acid, 99%
Supelco
Methylmalonic acid solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®