MilliporeSigma
  • Chemical cross-linking of cyclic AMP-dependent protein kinase and its dissimilar subunits.
Chemical cross-linking of cyclic AMP-dependent protein kinase and its dissimilar subunits.
The Biochemical journal (1983-03-01)
J P Charlton, C H Huang, L C Huang
ABSTRACT
Previous kinetic studies have demonstrated that the activation of cyclic AMP-dependent protein kinase by cyclic AMP involves the formation of a ternary complex of cyclic AMP, the regulatory subunit (R) and the catalytic subunit (C). It is suggested that only this ternary complex breaks down to liberate the enzymically active catalytic subunit. We have performed cross-linking experiments with the holoenzyme and its dissimilar subunits in the presence of MgATP and various concentrations of cyclic AMP. Results from these cross-linking studies indicate that regulatory subunits exist as dimers in the native form. Moreover, dissociation of the holoenzyme or the reconstituted enzyme is promoted by cyclic AMP, and the effect of MgATP is to stabilize the enzyme in the tetrameric form. The success in cross-linking the regulatory and the catalytic subunits of protein kinase with the lysine-specific bifunctional cross-linking reagent dimethyl suberimidate may be attributed to the presence of a large number of lysine residues in the enzyme.