MilliporeSigma
  • An insight into the regiospecificity of linoleic acid peroxidation catalyzed by mammalian 15-lipoxygenases.
An insight into the regiospecificity of linoleic acid peroxidation catalyzed by mammalian 15-lipoxygenases.
The journal of physical chemistry. B (2013-03-19)
Reynier Suardíaz, Laura Masgrau, José M Lluch, Angels González-Lafont
ABSTRACT
15-Lipoxygenases (15-LOs) catalyze the peroxidation reaction of linoleic acid (LA) in mammals producing almost exclusively 13-(S)-hydroperoxyoctadecadienoic acid (13-(S)-HPODE). Although several hypotheses have been formulated, the molecular basis of such enzymatic regiospecificity is unclear. We have here combined quantum mechanics/molecular mechanics (QM/MM) calculations with molecular dynamics simulations to analyze the peroxidation mechanism using a complete rabbit 15-LO-1/LA solvated model. C9 and C13 being equivalent for planarity and spin density, the QM/MM potential energy profiles of the O2 addition to those two atoms were calculated. The difference in the potential energy barrier heights is clear enough to justify that O2 selectively attacks C13 giving 13-(S)-HPODE. Oxygenation at C9 is hindered by two steric-shielding residues (Leu597 and Gln548). The calculated free energy profile at 300 K for the O2 addition to C13 confirms that the peroxidation on C13 is a reversible viable process in agreement with experiments. Thus, the subsequent reduction of the peroxyl radical to give the final hydroperoxidated product is expected to give the irreversibility character to the overall process.
Sigma-Aldrich
Linoleic acid sodium salt, ≥97% (GC)
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
α-Linoleic acid, ≥98%