MilliporeSigma
  • Prognosis of patients with methicillin-resistant Staphylococcus aureus bloodstream infection treated with teicoplanin: a retrospective cohort study investigating effect of teicoplanin minimum inhibitory concentrations.
Prognosis of patients with methicillin-resistant Staphylococcus aureus bloodstream infection treated with teicoplanin: a retrospective cohort study investigating effect of teicoplanin minimum inhibitory concentrations.
BMC infectious diseases (2013-04-23)
Jann-Tay Wang, Hau-Shin Wu, Chia-Min Weng, Le-Yin Hsu, Fu-Der Wang
ABSTRACT
The present study was designed to investigate whether teicoplanin minimum inhibitory concentrations (MICs) of methicillin-resistant Staphylococcus aureus (MRSA) isolates play a role in the prognosis of patient with teicoplanin-treated MRSA bloodstream infection (BSI). Between 1 January 2006 and 31 December 2009, adult patients with teicoplanin-treated MRSA BSI in two Taiwan medical centers were retrospectively enrolled. Their blood MRSA isolates were submitted for determination of MICs to various antibiotics and multi-locus sequence types. All-cause mortalities on Days 14 and 30, as well as clinical response at the end of teicoplanin therapy were treated as endpoints. Two hundred seventy adult patients were enrolled and 210 blood MRSA isolates were available. Independent risk factors for un-favorable outcome at the end of teicoplanin therapy included septic shock (p < 0.0001) and an elevated C-reactive protein level (p = 0.0064). The independent risk factors for all-cause Day 14 mortality (13.0%) included the presence of auto-immune diseases (p = 0.0235), septic shock (p = 0.0253) and thrombocytopenia (p = 0.0018). The independent risk factors for all-cause Day 30 mortality (26.3%) included age (p = 0.0102), septic shock (p < 0.0001) and thrombocytopenia (p = 0.0059). The current study didn't find a significant role for teicoplanin MICs in the prognosis of adult patients with teicoplanin-treated MRSA BSI.
Sigma-Aldrich
Teicoplanin
Pricing and availability is not currently available.