MilliporeSigma
[Pain in connection with colonoscopy in Norway].
Tidsskrift for den Norske laegeforening : tidsskrift for praktisk medicin, ny raekke (2013-05-29)
Øyvind Holme, Volker Moritz, Michael Bretthauer, Birgitte Seip, Tom Glomsaker, Thomas de Lange, Lars Aabakken, Asbjørn Stallemo, Ole Høie, Stein Dahler, Per Kristian Sandvei, Njaal Stray, Carl Magnus Ystrøm, Geir Hoff
ABSTRACT
Colonoscopies are common examinations at Norwegian hospitals. In contrast to many other countries, the majority of colonoscopies in Norway are conducted without routine sedation or analgesia. We wanted to investigate whether current Norwegian practice offers adequate pain relief. The material consists of prospectively recorded outpatient colonoscopies in the period January 2003-December 2011 performed at Norwegian hospitals in the quality assurance network for gastrointestinal endoscopy (Gastronet). We analysed demographic patient data and data from colonoscopies. Patients' experience of pain (none, slight, moderate or severe pain) in connection with the examination was established with the aid of a validated questionnaire. Data from 61,749 colonoscopies (55% on women) performed at 29 different hospitals were analysed. Colonoscopies were perceived as moderately or very painful by 33% of the patients (41% of the women, 24% of the men, p < 0.001). There were substantial differences between hospitals as to the percentage of colonoscopies that were perceived as moderately or very painful (from 9% to 43%, p < 0.001) and the use of sedatives and analgesics for the colonoscopies (from 1% to 92% of the examinations, p < 0.001). Only 23% of those who found the colonoscopy painful received analgesics. Pethidine was used in 95% of the cases in which analgesics were used during the examination. Many patients find colonoscopies painful. Pain relief practice varies substantially between hospitals. Pethidine is an analgesic with a slow onset of action, and should perhaps be replaced with more rapidly acting opiates.