MilliporeSigma
  • Adult duct-lining cells can reprogram into β-like cells able to counter repeated cycles of toxin-induced diabetes.
Adult duct-lining cells can reprogram into β-like cells able to counter repeated cycles of toxin-induced diabetes.
Developmental cell (2013-07-03)
Keith Al-Hasani, Anja Pfeifer, Monica Courtney, Nouha Ben-Othman, Elisabet Gjernes, Andhira Vieira, Noémie Druelle, Fabio Avolio, Philippe Ravassard, Gunter Leuckx, Sandra Lacas-Gervais, Damien Ambrosetti, Emmanuel Benizri, Jacob Hecksher-Sorensen, Pierre Gounon, Jorge Ferrer, Gerard Gradwohl, Harry Heimberg, Ahmed Mansouri, Patrick Collombat
ABSTRACT
It was recently demonstrated that embryonic glucagon-producing cells in the pancreas can regenerate and convert into insulin-producing β-like cells through the constitutive/ectopic expression of the Pax4 gene. However, whether α cells in adult mice display the same plasticity is unknown. Similarly, the mechanisms underlying such reprogramming remain unclear. We now demonstrate that the misexpression of Pax4 in glucagon(+) cells age-independently induces their conversion into β-like cells and their glucagon shortage-mediated replacement, resulting in islet hypertrophy and in an unexpected islet neogenesis. Combining several lineage-tracing approaches, we show that, upon Pax4-mediated α-to-β-like cell conversion, pancreatic duct-lining precursor cells are continuously mobilized, re-express the developmental gene Ngn3, and successively adopt a glucagon(+) and a β-like cell identity through a mechanism involving the reawakening of the epithelial-to-mesenchymal transition. Importantly, these processes can repeatedly regenerate the whole β cell mass and thereby reverse several rounds of toxin-induced diabetes, providing perspectives to design therapeutic regenerative strategies.
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder