MilliporeSigma
  • Compound I formation is a partially rate-limiting process in chloroperoxidase-catalyzed bromination reactions.
Compound I formation is a partially rate-limiting process in chloroperoxidase-catalyzed bromination reactions.
The Journal of biological chemistry (1990-09-05)
R D Libby, N S Rotberg
ABSTRACT
The kinetics of chloroperoxidase-catalyzed bromination and chlorination reactions were studied at various halide and hydrogen peroxide concentrations. At very high concentrations, both chloride (KI = 370 mM) and bromide (KI = 150 mM) are competitive substrate inhibitors versus hydrogen peroxide. Results at subinhibitory halide concentrations for bromination reactions (kcat = 4 ms-1, kcat/KPeroxide = 1.6 microM-1 x s-1 and kcat/KBr = 4.0 microM-1 x s-1) and chlorination reactions (kcat = 1.5 ms-1, kcat/Kperoxide = 2.3 microM-1 x s-1, and kcat/KBr = 0.32 microM-1 x s-1) indicate that halide oxidation is rate-limiting in chlorination reactions. However, in bromination reactions, both compound I formation and bromide oxidation are partially rate-limiting. This is the first documented case where compound I formation participates in determining the overall rate of a peroxidase reaction.