MilliporeSigma
  • Plasma 19-hydroxyandrostenedione and androstenedione levels in hydatidiform moles and early pregnancies with and without hypertensive diseases.
Plasma 19-hydroxyandrostenedione and androstenedione levels in hydatidiform moles and early pregnancies with and without hypertensive diseases.
The Australian & New Zealand journal of obstetrics & gynaecology (1989-08-01)
M E Hähnel, J D Martin, R Hähnel
ABSTRACT
Because of the association of preeclampsia with hydatidiform mole, and since levels of 19-hydroxyandrostenedione (19-OH-A) and androstenedione (A) are raised in hypertensive diseases of pregnancy, we compared plasma 19-OH-A and A in hydatidiform mole patients with control early pregnant women. Both steroids were significantly elevated in 27 patients with partial or complete hydatidiform moles, as compared with the levels in 36 control pregnancies at similar stages of gestation.