MilliporeSigma
  • Lateral diffusion of phospholipids in the lipid surface of human low-density lipoprotein measured with a pyrenyl phospholipid probe.
Lateral diffusion of phospholipids in the lipid surface of human low-density lipoprotein measured with a pyrenyl phospholipid probe.
European journal of biochemistry (1989-12-22)
M Vauhkonen, M Sassaroli, P Somerharju, J Eisinger
ABSTRACT
Human low-density lipoprotein (LDL) was labelled with the excimeric fluorescent phospholipid analogue 1-palmitoyl-2-(1'-pyreneoctanoyl)-sn-glycero-3-phosphocholine by using phosphatidylcholine-specific transfer protein for the probe insertion. The lateral diffusivity of the probe in the phospholipid/cholesterol surface monolayer of LDL was determined from the measured dependence of the pyrene monomer fluorescence yield on probe concentration. The data were analyzed by the milling-crowd model (J. Eisinger et al. (1986) Biophys. J. 49, 987-1001] to obtain the short-range lateral diffusivity of the probe. The lateral mobility of the probe in LDL was compared to that in model lipid systems, i.e. in protein-free LDL-like lipid particles and in small unilamellar vesicles, with a phospholipid/cholesterol composition characteristic of LDL. This analysis with the probability PE = 1 for excimer production between nearest-neighbour probes gives the lower limits for f, the frequency of translational lipid--lipid exchanges of the probe of 0.62 x 10(8), 0.19 x 10(8) and 0.19 x 10(8)s-1 in LDL, LDL-like lipid particles, and small unilamellar vesicles, respectively. The lower limits for the corresponding lateral diffusion constants are 16, 5 and 5 microns 2 s-1. The results suggest that the translational mobility of phospholipid molecules in the lipid--protein surface of LDL is not constrained by the apolipoprotein B-100 moiety or the neutral lipid core of the lipoprotein. Instead, the protein moiety may perturb the lipid order with the lipid--associating peptide domains and thus fluidize the amphiphilic surface monolayer of LDL relative to the protein-free model systems. In general, lateral diffusivity of the pyrenyl phospholipid probe in LDL and the model lipid systems is comparable to the lateral mobility of lipid analogue probes in a variety of model and biological membranes.