MilliporeSigma
  • Identification and quantification of p-hydroxyethylamphetamine as a novel metabolite of ethylamphetamine in rat by gas chromatography-mass spectrometry.
Identification and quantification of p-hydroxyethylamphetamine as a novel metabolite of ethylamphetamine in rat by gas chromatography-mass spectrometry.
Forensic science international (1989-04-01)
Y Makino, T Higuchi, S Ohta, M Hirobe
ABSTRACT
Ethylamphetamine and two major metabolites in urine, obtained after intraperitoneal administration of the drug to rats, have been isolated by gas chromatography-mass spectrometry. We have also synthesized an authentic sample of p-hydroxy-N-ethylamphetamine by a simple method. In rat, the metabolic pathway of ethylamphetamine is similar to that of amphetamine and methylamphetamine according to quantitative analysis of the major metabolites using a selected ion monitoring (SIM) method.