MilliporeSigma
  • An ESR study of nonenzymatic reactions of nitroso compounds with biological reducing agents.
An ESR study of nonenzymatic reactions of nitroso compounds with biological reducing agents.
Free radical research communications (1988-01-01)
N Takahashi, V Fischer, J Schreiber, R P Mason
ABSTRACT
The nonenzymatic reduction of nitrosobenzene (I), 2-nitroso-1-naphthol (II) and 2-nitroso-1-naphthol-4-sulfonic acid (III) with reducing agents such as NADPH, L-cysteine and N-acetyl-L-cysteine led to the formation of the corresponding hydronitroxide radicals, as confirmed with ESR spectroscopy. In addition to these radicals, a novel hydronitroxide radical, which was conjugated with GSH at the 4-position, was observed in the reaction of II or III with GSH. The formation of a hydronitroxide conjugated with GSH still retains the radical structure with its related redox chemistry. In this case, the formation of a GSH conjugate does not lead to the formation of chemically less reactive species.
Sigma-Aldrich
1-Nitroso-2-naphthol, 97%