MilliporeSigma
  • Effects of a thiazide diuretic (hydroflumethiazide) and a loop diuretic (bumetanide) on the endocrine pancreas: studies in vitro.
Effects of a thiazide diuretic (hydroflumethiazide) and a loop diuretic (bumetanide) on the endocrine pancreas: studies in vitro.
Metabolism: clinical and experimental (1985-08-01)
K Hermansen, O Schmitz, C E Mogensen
ABSTRACT
Treatment with thiazide diuretics causes an impairment of the glucose metabolism. To study whether this is due to a direct effect on the endocrine pancreas, the effects of the thiazide hydroflumethiazide on the release of glucagon, insulin, and somatostatin from the isolated perfused pancreas of normal and alloxan diabetic dogs were examined. Hydroflumethiazide at concentrations ranging from 1 to 50 micrograms/mL stimulated the normal secretion of glucagon (P less than 0.001), insulin (P less than 0.001), and somatostatin (P less than 0.001) in a dose-dependent manner. The normal hormone responses evoked by 50 micrograms/mL of the thiazide were, however, modified by the prevailing glucose level: higher insulin (P less than 0.05) and somatostatin (P less than 0.05) and lower glucagon (P less than 0.05) were obtained at the high glucose concentration of 11 mmol/L rather than at the low glucose concentration of 1.3 mmol/L. In alloxan diabetes, insulin secretion was almost extinct and did not respond to hydroflumethiazide, whereas glucagon was dose-dependently stimulated (P less than 0.001). In addition, we looked at the effect of the loop diuretic, bumetanide. The infusion of bumetanide at doses ranging from 0.5 to 3 micrograms/mL did not alter the release of glucagon, insulin, and somatostatin in the presence of 5.5 mmol/L glucose. The results suggest that hydroflumethiazide possesses the ability to directly stimulate A cell secretion in the normal and alloxan diabetic pancreas. Whether this effect is of clinical importance for the diminution in glucose tolerance observed during thiazide therapy remains, however, uncertain.