- Novel O-linked carbohydrate chains in the cellulase complex (cellulosome) of Clostridium thermocellum. 3-O-Methyl-N-acetylglucosamine as a constituent of a glycoprotein.
Novel O-linked carbohydrate chains in the cellulase complex (cellulosome) of Clostridium thermocellum. 3-O-Methyl-N-acetylglucosamine as a constituent of a glycoprotein.
The Journal of biological chemistry (1989-01-15)
G J Gerwig, P de Waard, J P Kamerling, J F Vliegenthart, E Morgenstern, R Lamed, E A Bayer
PMID2910842
ABSTRACT
Alkaline borohydride treatment of the cellulosome of Clostridium thermocellum yielded two major oligosaccharide-alditols, namely D-Galp-beta(1----4)-D-GalOH and (formula; see text) The compounds, isolated via gel permeation chromatography and high performance liquid chromatography, were analyzed by monosaccharide analysis, methylation analysis, gas-liquid chromatography/mass spectrometry, fast atom bombardment/mass spectrometry, and one- and two-dimensional 500-MHz (COSY, HOHAHA, ROESY) 1H NMR spectroscopy. Sodium dodecyl sulfate-polyacrylamide gel electrophoresis combined with blotting technology indicated that the tetrasaccharide is mainly associated with one of the cellulosome subunits.