MilliporeSigma
  • Influence of long-term treatment with hydrazinophthalazines on the activity of lysosomal exoglycosidases in rat tissues.
Influence of long-term treatment with hydrazinophthalazines on the activity of lysosomal exoglycosidases in rat tissues.
Acta medica Hungarica (1988-01-01)
K Olczyk, E J Kucharz, M Drózdz
ABSTRACT
Serum antinuclear antibodies were induced in rats by treatment with hydralazine or todralazine, given in daily dose of 25 mg/kg of body wt. for 12 months. An increased activity of alpha-glucosidase, beta-glucosidase, beta-galactosidase, alpha-fucosidase, beta-xylosidase was found in heart muscle, liver, kidney and lung. Hydralazine produced a more significant increase in the enzyme activity than todralazine. The obtained results suggest drug-induced connective tissue damage, probably of an inflammatory type. The mechanism of this phenomenon and its relationship to immunological alterations remain unclear.