MilliporeSigma
  • Inhibitor properties of some 5-substituted uracil acyclonucleosides, and 2,2'-anhydrouridines versus uridine phosphorylase from E. coli and mammalian sources.
Inhibitor properties of some 5-substituted uracil acyclonucleosides, and 2,2'-anhydrouridines versus uridine phosphorylase from E. coli and mammalian sources.
Biochemical pharmacology (1987-12-01)
A K Drabikowska, L Lissowska, Z Veres, D Shugar
ABSTRACT
Two series of 5-substituted uracil N(1)-acyclonucleosides, each with a different acyclic chain, were examined as inhibitors of uridine phosphorylase from rat intestinal mucosa, and several against the enzyme from Ehrlich ascites cells. In addition, several 5-substituted analogues of 2,2'-anhydrouridine were tested for their inhibitory effects vs a highly purified uridine phosphorylase from Escherichia coli. The results are compared with previously published data for inhibition of the E. coli enzyme by the acyclonucleosides, and of the rat enzyme by the anhydrouridines. In all instances, the inhibitors were active only vs the uridine, but not thymidine, phosphorylase from E. coli, and inhibition was competitive with respect to uridine as substrate. In general, with one or two exceptions, inhibitory effects were more pronounced against the enzyme from mammalian sources. Amongst the acyclonucleoside analogues, the most effective inhibitor of the enzyme from the rat and Ehrlich ascites cells exhibited a Ki = 0.1 microM, comparable to that reported with the Sarcoma-180 enzyme, whereas the Ki for inhibition of the E. coli enzyme was 0.7 microM. By contrast, another effective inhibitor of the bacterial enzyme was 7-fold less potent against the mammalian enzyme. The 2,2'-anhydrouridines were 10- to 30-fold more effective against the rat, as compared to the E. coli, enzyme. The overall quantitative data provide a reasonably good basis for the further design of potent inhibitors for possible use in chemotherapy.