MilliporeSigma
  • High-performance liquid chromatographic separation and electrochemical or spectrophotometric determination of R(-)N-n-propylnorapomorphine and R(-)10,11-methylenedioxy-N-n-propylnoraporphine in primate plasma.
High-performance liquid chromatographic separation and electrochemical or spectrophotometric determination of R(-)N-n-propylnorapomorphine and R(-)10,11-methylenedioxy-N-n-propylnoraporphine in primate plasma.
Journal of chromatography (1988-04-29)
P Lampen, J L Neumeyer, R J Baldessarini
ABSTRACT
The dopamine receptor agonist R(-)N-n-propylnorapomorphine (NPA) and its proposed pro-drug R(-)10,11-methylenedioxy-N-n-propylnoraporphine (MDO-NPA) were isolated simultaneously from monkey plasma using a solid-phase extraction procedure. R(-)Apomorphine (APO) and R(-)10,11-methylenedioxyaporphine (MDO-APO) were added as internal standards, and separation and quantification were by high-performance liquid chromatography with electrochemical or ultraviolet detection of the free catechol and MDO compounds, respectively. The detection limits for NPA and MDO-NPA in plasma were 0.5 and 10 ng/ml and the coefficient of variation (S.D./mean) within assays and between days of assays for both drugs was 5.6% or less. Quantification of plasma levels of NPA and MDO-NPA was possible at ranges of 2-1000 and 40-5000 ng/ml, respectively, including concentrations found after intravenous administration of these agents.