MilliporeSigma
  • [The nitrazine yellow test. A simple clinical method for diagnosing epidermal wound healing].
[The nitrazine yellow test. A simple clinical method for diagnosing epidermal wound healing].
Zeitschrift fur Hautkrankheiten (1987-08-01)
R Niedner, W Vanscheidt, H Wokalek
ABSTRACT
Nitrazine yellow test has proved successful in determining toxic degenerative hand eczema. Additionally, it may be regarded as helpful in the quantitative registration of kinetic data on epidermal wound healing.