MilliporeSigma
  • Glutathione S-transferase in adult Dirofilaria immitis and Brugia pahangi.
Glutathione S-transferase in adult Dirofilaria immitis and Brugia pahangi.
Molecular and biochemical parasitology (1986-08-01)
J J Jaffe, R A Lambert
ABSTRACT
Glutathione S-transferase (EC 2.5.1.18) was detected in the cytosolic and microsomal fractions of adult Dirofilaria immitis females at respective levels of 30 nmol and 3 nmol min-1 (mg protein)-1 activity with the substrate 1-chloro-2,4-dinitrobenzene (CDNB). The transferase activity in the cytosolic fraction of adult Brugia pahangi females was 10 nmol min-1 mg-1 with CDNB; determination of its activity in the microsomal fraction of this filariid was not attempted. These filarial glutathione S-transferases were further characterized after their purification by glutathione-affinity chromatography. Sodium dodecyl sulfate-polyacrylamide gel electrophoresis of the cytosolic transferase from D. immitis, molecular weight 47000, yielded a single subunit of around 28 kDa. The cytosolic and microsomal transferases from D. immitis differed in their activity with CDNB, 1,2-dichloro-4-nitrobenzene, 4-benzylchloride and ethacrynic acid. The cytosolic transferase from B. pahangi was distinguished by its high activity with ethacrynic acid. Both glutathione S-transferases from D. immitis also functioned as a glutathione peroxidase, strongly preferring cumene hydroperoxide as a substrate over hydrogen peroxide. Both were equiactive inhibitors of malonaldehyde formation in the NADPH-microsomal lipid peroxidation system. Thus, in addition to the ability of filarial glutathione S-transferases to detoxify electrophilic xenobiotics, at least those from D. immitis also exhibited selenium-independent glutathione peroxidase activity. Their glutathione S-transferase function suggests a potential role for these enzymes in the leukotriene synthetic pathway, if filariae can form such eicosanoids from arachidonate. Functioning as a glutathione peroxidase, they could serve to protect filarial membrane lipids from peroxidation.
Sigma-Aldrich
4-Nitrobenzyl chloride, 99%