MilliporeSigma
  • Antiviral and interferon-inducing properties of interferon inducers administered with prostaglandins.
Antiviral and interferon-inducing properties of interferon inducers administered with prostaglandins.
Antimicrobial agents and chemotherapy (1980-03-01)
D A Stringfellow
ABSTRACT
Prostaglandins enhanced the interferon response and therapeutic activity of interferon inducers in Friend leukemia virus-infected mice. A similar enhancement in interfron responsiveness but not antiviral activity was observed in mice infected with rapidly acute viruses.