MilliporeSigma
  • Effects of a TRH analog (DN-1417) and TRH-T on the electroconvulsive threshold.
Effects of a TRH analog (DN-1417) and TRH-T on the electroconvulsive threshold.
The Kurume medical journal (1983-01-01)
J Nakamura, N Uchimura, S Yamada, T Tsutsumi, K Ishibashi, H Kojima, K Inanaga