MilliporeSigma
The neurotoxins of the sea snake Laticauda schistorhynchus.
The Biochemical journal (1983-07-01)
M L Guinea, N Tamiya, H G Cogger
ABSTRACT
Erabutoxins a and b, the major neurotoxins in the venom of the sea snake Laticauda semifasciata, were detected in the venom of Laticauda schistorhynchus. The identity of the toxins was confirmed on the basis of elution position on CM-cellulose column chromatography, disc electrophoretic mobility, amino acid analysis and toxicity measurement.