MilliporeSigma
  • Bile acid inhibition of basic and neutral glutathione S-transferases in rat liver.
Bile acid inhibition of basic and neutral glutathione S-transferases in rat liver.
The Biochemical journal (1983-12-01)
J D Hayes, J Chalmers
ABSTRACT
A purification scheme is described for the neutral glutathione S-transferases of rat liver. Discontinuous sodium dodecyl sulphate/polyacrylamide-gel electrophoresis revealed that one of these enzymes contains a previously unidentified subunit, which has a molecular mass of 23 000 Da and has been designated Yn. Bile acids inhibited the activity of all the basic and neutral transferases investigated, but marked differences in the effects of bile acids on individual enzymes were observed. The activity of each transferase was inhibited more by lithocholate 3-sulphate than by chenodeoxycholate, which in turn was more inhibitory than cholate. The enzymes that were most sensitive to cholate inhibition were not found to be as readily inhibited as other transferases by chenodeoxycholate or lithocholate 3-sulphate. Conversely, the activity of transferase AA was more resistant to cholate, chenodeoxycholate and lithocholate 3-sulphate inhibition than was any of the other enzymes studied.