MilliporeSigma
[Modification of the recovery of reserpine-induced amine depletion].
Biomedica biochimica acta (1983-01-01)
K F Funk, J Schmidt
ABSTRACT
Recovery of the dopamine storage pool after depletion by reserpine was slightly enhanced by treatment with nandrolone but remained unaffected by nootropic drugs.