MilliporeSigma
  • Cinromide's metabolite in monkey model: gastric administration and seizure control.
Cinromide's metabolite in monkey model: gastric administration and seizure control.
Epilepsia (1980-04-01)
J S Lockard, R H Levy, L L DuCharme, W C Congdon
ABSTRACT
In a previous study (Lockard et al., 1979) Cinromide (3 bromo-N-ethylcinnamamide), an experimental anticonvulsant (Burroughs-Wellcome Pharmaceutical Co.), was given a preliminary evaluation. Since that research was concerned primarily with EEG paroxysms, the present study was conducted to address drug efficacy in terms of clinical seizures. Cinromide's major metabolite (3-bromocinnamamide, BC) was the main focus. Eight alumina-gel monkeys were given by gastric bolus every 6 hr for 10 days (Phase I) either the solvent alone (Tween 80), Cinromide (BEC), or its synthetic metabolite (SBC). Subsequently (Phase II), four animals were given BEC or SBC by chronic gastric infusion for 20 days. In both phases Cinromide's metabolite (either via BEC or especially SBC) was effective in half of the animals in reducing seizure frequency and/or duration at plasma levels above 5 mcg/m. The data suggest that the drug's efficacy is individually specific. Another species of Cinromide metabolism, 3-bromocinnamic acid, is also discussed.