MilliporeSigma
  • Prolidase deficiency with iminodipeptiduria: biochemical investigations and first results of attempted therapy.
Prolidase deficiency with iminodipeptiduria: biochemical investigations and first results of attempted therapy.
Journal of inherited metabolic disease (1981-01-01)
C Charpentier, K Dagbovie, A Lemonnier, M Larregue, R A Johnstone
ABSTRACT
A 33-year-old female patient with chronic recurrent leg ulcerations was shown to present a massive iminodipeptiduria which seemed to be attributable to disturbance of collagen metabolism. Biochemical investigations confirmed an hereditary prolidase deficiency. A treatment was tried for the first time and showed a good biochemical result and a clinical improvement.
Sigma-Aldrich
trans-1-Acetyl-4-hydroxy-L-proline, ≥98%