MilliporeSigma
  • Comparative tissue absorption of oral 14C-aspirin and topical triethanolamine 14C-salicylate in human and canine knee joints.
Comparative tissue absorption of oral 14C-aspirin and topical triethanolamine 14C-salicylate in human and canine knee joints.
Journal of clinical pharmacology (1982-01-01)
J L Rabinowitz, E S Feldman, A Weinberger, H R Schumacher
ABSTRACT
The local, articular, and systemic absorption of oral and topical salicylates was studied in dogs and humans using radioisotope techniques. Topical triethanolamine 14C-salicylate was found capable of percutaneous absorption into the knee joint and surrounding tissues. In dogs, topical salicylate application resulted in higher salicylate concentrations than oral aspirin in a number of tissues, despite lower blood levels. In patients with rheumatoid arthritis, intraarticular 14C-salicylate levels after triethanolamine 14C-salicylate cream were 60 per cent of those obtained with oral aspirin. Four of six patients reported equal improvement in local discomfort after oral and topical salicylates. A potential role for topical salicylate cream in the treatment of localized rheumatic disorders is suggested.