MilliporeSigma
  • Acid-labile resin linkage agents for use in solid phase peptide synthesis.
Acid-labile resin linkage agents for use in solid phase peptide synthesis.
International journal of peptide and protein research (1982-11-01)
R C Sheppard, B J Williams
ABSTRACT
Details are given of the preparation of two acid-labile peptide-resin linkage agents for use in solid phase peptide synthesis, viz. 4-hydroxymethylphenoxy-acetic acid and 3-methoxy-4-hydroxymethylphenoxyacetic acid. The latter is suitable for the preparation of peptide fragments bearing t-butyl-based side chain protecting groups.