MilliporeSigma
  • A swine dysentery model for evaluation of drug prophylaxis: efficacy of various drugs in the control of swine dysentery.
A swine dysentery model for evaluation of drug prophylaxis: efficacy of various drugs in the control of swine dysentery.
American journal of veterinary research (1981-01-01)
J P Raynaud, G Brunault, E B Patterson
ABSTRACT
A swine dysentery (SD) model that produces consistent, homogeneous, and severe SD was used in 2 experiments to compare the prophylactic effectiveness of 5 commercially available swine feed additive products. Under the conditions of these studies, carbadox and carbadox + sulfamethazine proved to be the most effective agents in preventing SD during the infection + medication and postmedication periods. Olaquindox was effective in preventing SD in the infection + medication period; however, SD recurrence was high during the postmedication period. Nithiamide and chlortetracycline + sulfamethazine + penicillin were least effective in preventing SD during the infection + medication and postmedication periods.