MilliporeSigma
  • [Biopharmaceutical studies of drugs. III. Serum concentration level of o-ethoxybenzamide and N-(beta-hydroxybutyryl)-p-phenetidine after simultaneous administration to rabbit (author's transl)].
[Biopharmaceutical studies of drugs. III. Serum concentration level of o-ethoxybenzamide and N-(beta-hydroxybutyryl)-p-phenetidine after simultaneous administration to rabbit (author's transl)].
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan (1981-06-01)
K Kigasawa, M Hiiragi, T Hayashi