MilliporeSigma
  • Stress response of juvenile sockeye salmon (Oncorhynchus nerka) to the butoxyethanol ester of 2,4-dichlorophenoxyacetic acid.
Stress response of juvenile sockeye salmon (Oncorhynchus nerka) to the butoxyethanol ester of 2,4-dichlorophenoxyacetic acid.
Bulletin of environmental contamination and toxicology (1981-12-01)
J R McBride, H M Dye, E M Donaldson