MilliporeSigma
  • Synthesis and sigma binding properties of 1'- and 3'-halo- and 1',3'-dihalo-N-normetazocine analogues.
Synthesis and sigma binding properties of 1'- and 3'-halo- and 1',3'-dihalo-N-normetazocine analogues.
Journal of medicinal chemistry (1995-07-21)
R Danso-Danquah, X Bai, X Zhang, S W Mascarella, W Williams, B Sine, W D Bowen, F I Carroll
ABSTRACT
The synthesis and sigma 1 and sigma 2 binding properties of several 1'- and 3'-halo- and 1',3'-dihalo-substituted analogues of (+)-N-benzyl- and (+)- and (-)-N-dimethylallyl-N-normetazocine are presented. Structure-activity relationship analyses of the binding data showed that halogen substitution at the 1'-position of these N-substituted N-normetazocine analogues had little effect on sigma 1 binding affinity, whereas 3'-halo substitution as well as 1',3'-dihalo substitution resulted in a reduction of affinity. sigma 2 affinity was increased by the presence of a 3'-bromo substituent in this series of (+)-N-substituted N-normetazocines.