MilliporeSigma
  • SPECT imaging of dopamine and serotonin transporters with [123I]beta-CIT: pharmacological characterization of brain uptake in nonhuman primates.
SPECT imaging of dopamine and serotonin transporters with [123I]beta-CIT: pharmacological characterization of brain uptake in nonhuman primates.
Synapse (New York, N.Y.) (1993-04-01)
M Laruelle, R M Baldwin, R T Malison, Y Zea-Ponce, S S Zoghbi, M S al-Tikriti, E H Sybirska, R C Zimmermann, G Wisniewski, J L Neumeyer
ABSTRACT
Single photon emission computed tomography (SPECT) studies of regional kinetic uptake and pharmacological specificity of [123I]methyl 3 beta-(4-iodophenyl) tropane-2 beta-carboxylate ([123I]beta-CIT) were performed in nonhuman primates (n = 41). In control experiments, activity was concentrated in striatum and in hypothalamic/midbrain regions. Striatal uptake increased for 140-180 min and displayed stable levels thereafter. Striatal to cerebellar activity ratios were 7.3 +/- 0.9 (mean +/- SEM) at 300 min. About 75% of striatal uptake was displaceable by injection of nonradioactive beta-CIT. Hypothalamic/midbrain activity reached maximal levels at approximately 45 min. A slow washout phase followed this peak activity. Activities in frontal, occipital, and cerebellar regions were characterized by an early peak (20-30 min), followed by rapid washout. Displacement studies demonstrated that striatal uptake was associated with dopamine (DA) transporters, as it was displaced by GBR 12909, a selective DA uptake inhibitor, but not by citalopram, a selective serotonin (5-HT) uptake inhibitor. The inverse was true in the hypothalamic/midbrain area, suggesting that the uptake in this area was associated primarily with 5-HT transporters. Maprotiline, a selective norepinephrine uptake inhibitor, did not affect [123I]beta-CIT uptake. In vivo site occupancy ED50 values of cocaine, 2 beta-carbomethoxy-3 beta-(4-fluorophenyl)tropane (CFT), and beta-CIT were measured in the striatum with a stepwise displacement paradigm. In vivo ED50 values correlated strongly with in vitro IC50 values for binding to DA transporters. Infusion of high dose of L-DOPA (250 mumol/kg) failed to displace striatal [123I]beta-CIT binding, suggesting that the binding would not be affected by L-DOPA administration in Parkinsonian patients. However, studies performed with injection of d-amphetamine indirectly suggested that high synaptic levels of DA may compete with [123I]beta-CIT binding. These studies suggest that [123I]beta-CIT will be a useful SPECT tracer of DA and 5-HT transporters in living human brain.
Sigma-Aldrich
Indatraline hydrochloride, solid